Please login to your online account to display your discounted pricing

Rat anti-CD44, Clone: IM7, Cy5, R-PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP141266PC5100TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP141266PC

  • / Each
 

Description & Specifications

Specifications

Formulation PBS [pH 7.4]
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 960
Antigen CD44
Host Species Rat
Immunogen Dexamethasone-induced cells of the SJL mouse spontaneous myeloid leukemia M1
Isotype IgG2b
Purification Method Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Protein A purified
Clone IM7
Conjugate Cy5, R-PE
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Storage Requirements Store at 4°C in the dark.
Test Specificity The rat monoclonal antibody IM7 reacts with CD44 antigen (Phagocyte glycoprotein 1), an 80-95kDa transmembrane glycoprotein (hyaladherin family) present on the most of cells and tissues (leukocytes, endothelial cells, mesenchymal cells, etc.); it is negative on platelets and hepatocytes. The antibody reacts with all isoforms of mouse CD44.
Concentration 1mg/mL