Please login to your online account to display your discounted pricing

Rat anti-CD43/Sialophorin, Clone: R2/60, FITC, Novus Biologicals™

$175.00 - $549.00

Specifications

Antigen CD43/Sialophorin
Clone R2/60
Immunogen The immunogen for this antibody was CD43.
Gene ID (Entrez) 6693
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14401101

 
novus biologicals canada
NBP14401101MG
0.1mg Each for $252.00

NBP14401105

 
novus biologicals canada
NBP144011005MG
0.05mg Each for $175.00

NBP1440115

 
novus biologicals canada
NBP14401105MG
0.5mg Each for $549.00
Description & Specifications

Specifications

Antigen CD43/Sialophorin
Clone R2/60
Immunogen The immunogen for this antibody was CD43.
Gene ID (Entrez) 6693
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgM
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry