Please login to your online account to display your discounted pricing

Rat anti-CD4, Clone: GK1.5, Azide Free, Novus Biologicals™

$300.00 - $300.00

Specifications

Antigen CD4
Clone GK1.5
Concentration 1mg/mL
Immunogen Mouse CTL clone V4 cells.
Gene ID (Entrez) 12504
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP237726

 
novus biologicals canada
NBP237726
0.1mg Each for $300.00

NBP237730

 
novus biologicals canada
NBP237730
0.1mg Each for $300.00
Description & Specifications

Specifications

Antigen CD4
Clone GK1.5
Concentration 1mg/mL
Immunogen Mouse CTL clone V4 cells.
Gene ID (Entrez) 12504
Formulation PBS. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Blocking
Applications Cell Depletion
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Neutralization