Please login to your online account to display your discounted pricing

Rat anti-CD4, Clone: GK1.5, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP237730

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP237730

  • / Each
 

Description & Specifications

Specifications

Formulation PBS. Azide Free.
Storage Requirements Store at 4°C. Do not freeze.
Antigen CD4
Gene ID (Entrez) 12504
Primary or Secondary Primary
Cross Reactivity Mouse
Applications Blocking
Applications Cell Depletion
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Neutralization
Test Specificity Antibody reacts with an extracellular epitope of mouse CD4 transmembrane glycoprotein (55kDa).
Immunogen Mouse CTL clone V4 cells.
Monoclonal or Polyclonal Monoclonal
Format Purified
Conjugate Unlabeled
Reconstitution Protein A purified
Clone GK1.5
Host Species Rat
Isotype IgG2b
Regulatory Status RUO
Purification Method Purified
Concentration 1mg/mL
Quantity 0.1mg