Please login to your online account to display your discounted pricing

Rat anti-CD4, Clone: GK1.5, Azide Free, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP237726

 View all options for this product


For Research Use Only

Catalog No. NBP237726


Description & Specifications


Antigen CD4
Applications Blocking
Applications Cell Depletion
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Neutralization
Clone GK1.5
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Purified
Formulation PBS. Azide Free.
Host Species Rat
Immunogen Mouse CTL clone V4 cells.
Isotype IgG2b
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 12504
Test Specificity Antibody reacts with an extracellular epitope of mouse CD4 transmembrane glycoprotein (55kDa).
Reconstitution Protein A purified