Please login to your online account to display your discounted pricing

Rat anti-CD36/SR-B3, Clone: No.72-1, PE, Novus Biologicals™

$192.00 - $289.00

Specifications

Antigen CD36/SR-B3
Clone No.72-1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD36.
Gene ID (Entrez) 948
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14387001

 
novus biologicals canada
NBP14387001MG
0.1mg Each for $289.00

NBP14387005

 
novus biologicals canada
NBP143870005MG
0.05mg Each for $192.00
Description & Specifications

Specifications

Antigen CD36/SR-B3
Clone No.72-1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD36.
Gene ID (Entrez) 948
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry