Please login to your online account to display your discounted pricing

Rat anti-CD36/SR-B3, Clone: No.72-1, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143870005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14387005

  • / Each
 

Description & Specifications

Specifications

Formulation PBS [pH 7.2]
Immunogen The immunogen for this antibody was CD36.
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Clone No.72-1
Cross Reactivity Mouse
Host Species Rat
Isotype IgG2a, κ
Quantity 0.05mg
Gene ID (Entrez) 948
Antigen CD36/SR-B3
Format Affinity Purified
Applications Flow Cytometry
Purification Method Affinity Purified
Primary or Secondary Primary
Concentration 0.2mg/mL
Conjugate PE