Please login to your online account to display your discounted pricing

Rat anti-CD27 Ligand/TNFSF7/CD70, Clone: FR70, PE, Novus Biologicals™

$329.00 - $472.00

Specifications

Antigen CD27 Ligand/TNFSF7/CD70
Clone FR70
Concentration 0.2mg/mL
Immunogen This antibody was generated by immunizing host with BALB/c mouse B lymphoma A20.2J
Gene ID (Entrez) 970
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20029626

 
novus biologicals canada
NBP200296200UG
200μg Each for $472.00

NBP20029610

 
novus biologicals canada
NBP200296100UG
100μg Each for $329.00
Description & Specifications

Specifications

Antigen CD27 Ligand/TNFSF7/CD70
Clone FR70
Concentration 0.2mg/mL
Immunogen This antibody was generated by immunizing host with BALB/c mouse B lymphoma A20.2J
Gene ID (Entrez) 970
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2b, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry