Please login to your online account to display your discounted pricing

Rat anti-CD27 Ligand/TNFSF7/CD70, Clone: FR70, Biotin, Novus Biologicals™

$131.00 - $395.00

Specifications

Antigen CD27 Ligand/TNFSF7/CD70
Clone FR70
Immunogen This antibody was generated by immunizing host with BALB/c mouse B lymphoma A20.2J
Gene ID (Entrez) 970
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14362005

 
novus biologicals canada
NBP143620005MG
0.05mg Each for $131.00

NBP14362001

 
novus biologicals canada
NBP14362001MG
0.1mg Each for $197.00

NBP1436205

 
novus biologicals canada
NBP14362005MG
0.5mg Each for $395.00
Description & Specifications

Specifications

Antigen CD27 Ligand/TNFSF7/CD70
Clone FR70
Immunogen This antibody was generated by immunizing host with BALB/c mouse B lymphoma A20.2J
Gene ID (Entrez) 970
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2b, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry