Please login to your online account to display your discounted pricing

Rat anti-CD25/IL-2 R alpha, Clone: 7D4, Biotin, Novus Biologicals™

$175.00 - $549.00

Specifications

Antigen CD25/IL-2 R alpha
Clone 7D4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD25.
Gene ID (Entrez) 3559
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14366201

 
novus biologicals canada
NBP14366201MG
0.1mg Each for $252.00

NBP14366205

 
novus biologicals canada
NBP143662005MG
0.05mg Each for $175.00

NBP1436625

 
novus biologicals canada
NBP14366205MG
0.5mg Each for $549.00
Description & Specifications

Specifications

Antigen CD25/IL-2 R alpha
Clone 7D4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD25.
Gene ID (Entrez) 3559
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgM, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry