Please login to your online account to display your discounted pricing

Rat anti-CD24 (p Ser123, p Ser124, p ser123, p ser124), Clone: 30-F1, FITC, Novus Biologicals™

$192.00 - $418.00

Specifications

Antigen CD24 (p Ser123, p Ser124, p ser123, p ser124)
Clone 30-F1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is CD24.
Gene ID (Entrez) 100133941
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20001010

 
novus biologicals canada
NBP200010100UG
100μg Each for $192.00

NBP20001055

 
novus biologicals canada
NBP200010500UG
500μg Each for $418.00
Description & Specifications

Specifications

Antigen CD24 (p Ser123, p Ser124, p ser123, p ser124)
Clone 30-F1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is CD24.
Gene ID (Entrez) 100133941
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2c, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry