Please login to your online account to display your discounted pricing

Rat anti-CD24, Clone: M1/69, FITC, Novus Biologicals™

$192.00 - $418.00

Specifications

Antigen CD24
Clone M1/69
Concentration 0.5mg/mL
Immunogen C57BL/10 mouse splenic T cells and concanavalin A-activated splenocytes
Gene ID (Entrez) 100133941
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20000710

 
novus biologicals canada
NBP200007100UG
100μg Each for $192.00

NBP20000753

 
novus biologicals canada
NBP200007500UG
500μg Each for $418.00
Description & Specifications

Specifications

Antigen CD24
Clone M1/69
Concentration 0.5mg/mL
Immunogen C57BL/10 mouse splenic T cells and concanavalin A-activated splenocytes
Gene ID (Entrez) 100133941
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry