Please login to your online account to display your discounted pricing

Rat anti-CD24, Clone: M1/69, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200007100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20000710

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Format Affinity Purified
Host Species Rat
Immunogen C57BL/10 mouse splenic T cells and concanavalin A-activated splenocytes
Isotype IgG2b, κ
Quantity 100μg
Regulatory Status RUO
Primary or Secondary Primary
Antigen CD24
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 100133941
Test Specificity Human reactivity reported in scientific literature (PMID: 24277698)
Clone M1/69
Concentration 0.5mg/mL
Conjugate FITC
Cross Reactivity Human
Cross Reactivity Mouse