Please login to your online account to display your discounted pricing

Rat anti-CD24, Clone: M1/69, Biotin, Novus Biologicals™

$128.00 - $386.00

Specifications

Antigen CD24
Clone M1/69
Concentration 0.5mg/mL
Immunogen C57BL/10 mouse splenic T cells and concanavalin A-activated splenocytes
Gene ID (Entrez) 100133941
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14366505

 
novus biologicals canada
NBP143665005MG
0.05mg Each for $128.00

NBP14366501

 
novus biologicals canada
NBP14366501MG
0.1mg Each for $192.00

NBP1436655

 
novus biologicals canada
NBP14366505MG
0.5mg Each for $386.00
Description & Specifications

Specifications

Antigen CD24
Clone M1/69
Concentration 0.5mg/mL
Immunogen C57BL/10 mouse splenic T cells and concanavalin A-activated splenocytes
Gene ID (Entrez) 100133941
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry