Please login to your online account to display your discounted pricing

Rat anti-CD24, Clone: M1/69, Biotin, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP14366505MG

 View all options for this product


For Research Use Only

Catalog No. NBP1436655


Description & Specifications


Antigen CD24
Applications Flow Cytometry
Clone M1/69
Concentration 0.5mg/mL
Conjugate Biotin
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen C57BL/10 mouse splenic T cells and concanavalin A-activated splenocytes
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 0.5mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 100133941
Test Specificity Human reactivity reported in scientific literature (PMID: 24277698)