Please login to your online account to display your discounted pricing

Rat anti-CD24, Clone: M1/69, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143665005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14366505

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Quantity 0.05mg
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 100133941
Conjugate Biotin
Cross Reactivity Human
Cross Reactivity Mouse
Antigen CD24
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen C57BL/10 mouse splenic T cells and concanavalin A-activated splenocytes
Isotype IgG2b, κ
Purification Method Affinity Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Test Specificity Human reactivity reported in scientific literature (PMID: 24277698)
Clone M1/69
Concentration 0.5mg/mL
Applications Flow Cytometry