Please login to your online account to display your discounted pricing

Rat anti-CD24, Clone: M1/69, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14366501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14366501

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Host Species Rat
Isotype IgG2b, κ
Antigen CD24
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Applications Flow Cytometry
Immunogen C57BL/10 mouse splenic T cells and concanavalin A-activated splenocytes
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 100133941
Clone M1/69
Concentration 0.5mg/mL
Conjugate Biotin
Cross Reactivity Human
Cross Reactivity Mouse
Purification Method Affinity Purified
Test Specificity Human reactivity reported in scientific literature (PMID: 24277698)