Please login to your online account to display your discounted pricing

Rat anti-CD23/Fc epsilon RII, Clone: B3B4, Biotin, Novus Biologicals™

$175.00 - $582.00

Specifications

Antigen CD23/Fc epsilon RII
Clone B3B4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD23.
Gene ID (Entrez) 2208
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14366601

 
novus biologicals canada
NBP14366601MG
0.1mg Each for $252.00

NBP14366605

 
novus biologicals canada
NBP143666005MG
0.05mg Each for $175.00

NBP1436665

 
novus biologicals canada
NBP14366605MG
0.5mg Each for $582.00
Description & Specifications

Specifications

Antigen CD23/Fc epsilon RII
Clone B3B4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD23.
Gene ID (Entrez) 2208
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry