Please login to your online account to display your discounted pricing

Rat anti-CD2, Clone: RM2-5, Biotin, Novus Biologicals™

$175.00 - $428.00

Specifications

Antigen CD2
Clone RM2-5
Concentration 0.5mg/mL
Immunogen BALB/c mouse thymocytes
Gene ID (Entrez) 914
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14369501

 
novus biologicals canada
NBP14369501MG
0.1mg Each for $197.00

NBP14369505

 
novus biologicals canada
NBP143695005MG
0.05mg Each for $175.00

NBP1436955

 
novus biologicals canada
NBP14369505MG
0.5mg Each for $428.00
Description & Specifications

Specifications

Antigen CD2
Clone RM2-5
Concentration 0.5mg/mL
Immunogen BALB/c mouse thymocytes
Gene ID (Entrez) 914
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2b, λ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry