missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-CD1d, Clone: 1B1, Novus Biologicals™

$219.00 - $406.00

Specifications

Antigen CD1d
Clone 1B1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD1d.
Gene ID (Entrez) 912
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14346101 Novus Biologicals Canada
NBP14346101MG
0.1mg Each for $274.00
NBP14346105 Novus Biologicals Canada
NBP143461005MG
0.05mg Each for $219.00
NBP1434615 Novus Biologicals Canada
NBP14346105MG
0.5mg Each for $406.00
Specifications Description & Specifications

Specifications

CD1d
0.5mg/mL
912
Monoclonal
Store at 4°C. Do not freeze.
IgG2b, κ
Affinity Purified
Flow Cytometry,Functional Assay,Immunohistochemistry,Immunohistochemistry (Frozen),Immunoprecipitation
1B1
The immunogen for this antibody was CD1d.
Phosphate buffer pH 7.2
Primary
Rat
Unlabeled
Affinity Purified