Please login to your online account to display your discounted pricing

Rat anti-CD1d, Clone: 1B1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143461005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14346105

  • / Each
 

Description & Specifications

Specifications

Antigen CD1d
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Immunogen The immunogen for this antibody was CD1d.
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 912
Clone 1B1
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse