missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-CD1d, Clone: 1B1, Biotin, Novus Biologicals™

$197.00 - $395.00

Specifications

Antigen CD1d
Clone 1B1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD1d.
Gene ID (Entrez) 912
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14369701 Novus Biologicals Canada
NBP14369701MG
0.1mg Each for $197.00
NBP1436975 Novus Biologicals Canada
NBP14369705MG
0.5mg Each for $395.00
Specifications Description & Specifications

Specifications

CD1d
0.2mg/mL
912
Monoclonal
Store at 4°C. Do not freeze.
IgG2b, κ
Affinity Purified
Flow Cytometry
1B1
The immunogen for this antibody was CD1d.
Phosphate buffer pH 7.2
Primary
Rat
Biotin
Affinity Purified