Please login to your online account to display your discounted pricing

Rat anti-CD160, Clone: CNX46-3, Novus Biologicals™

$307.00 - $384.00

Specifications

Antigen CD160
Clone CNX46-3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is CD160.
Gene ID (Entrez) 11126
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20019210

 
novus biologicals canada
NBP200192100UG
100μg Each for $307.00

NBP20019255

 
novus biologicals canada
NBP200192500UG
500μg Each for $384.00
Description & Specifications

Specifications

Antigen CD160
Clone CNX46-3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is CD160.
Gene ID (Entrez) 11126
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting