Please login to your online account to display your discounted pricing

Rat anti-CD160, Clone: CNX46-3, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP200192500UG

 View all options for this product


For Research Use Only

Catalog No. NBP20019255


Description & Specifications


Antigen CD160
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Clone CNX46-3
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Immunogen The immunogen for this antibody is CD160.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 500μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 11126