Please login to your online account to display your discounted pricing

Rat anti-CD160, Clone: CNX46-3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200192100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20019210

  • / Each
 

Description & Specifications

Specifications

Antigen CD160
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Clone CNX46-3
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Immunogen The immunogen for this antibody is CD160.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 100μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 11126