Please login to your online account to display your discounted pricing

Rat anti-CD16/CD32, Clone: 93, Biotin, Novus Biologicals™

$171.00 - $493.00

Specifications

Antigen CD16/CD32
Clone 93
Concentration 0.2mg/mL
Immunogen Murine CD16/32 (CD16/Fc gamma III and CD32/FcgammaII receptors)
Gene ID (Entrez) 2214
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14367201

 
novus biologicals canada
NBP14367201MG
0.1mg Each for $192.00

NBP14367205

 
novus biologicals canada
NBP143672005MG
0.05mg Each for $171.00

NBP1436725

 
novus biologicals canada
NBP14367205MG
0.5mg Each for $493.00
Description & Specifications

Specifications

Antigen CD16/CD32
Clone 93
Concentration 0.2mg/mL
Immunogen Murine CD16/32 (CD16/Fc gamma III and CD32/FcgammaII receptors)
Gene ID (Entrez) 2214
Formulation Phosphate buffer pH 7.2. IgG purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a
Conjugate Biotin
Format Purified
Purification Method Purified
Applications Flow Cytometry