Please login to your online account to display your discounted pricing

Rat anti-CD16/CD32, Clone: 93, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14367205MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1436725

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Regulatory Status RUO
Host Species Rat
Storage Requirements Store at 4°C. Do not freeze.
Immunogen Murine CD16/32 (CD16/Fc gamma III and CD32/FcgammaII receptors)
Conjugate Biotin
Concentration 0.2mg/mL
Antigen CD16/CD32
Purification Method Purified
Quantity 0.5mg
Isotype IgG2a
Primary or Secondary Primary
Clone 93
Format Purified
Gene ID (Entrez) 2214
Formulation Phosphate buffer pH 7.2. IgG purified.
Monoclonal or Polyclonal Monoclonal
Test Specificity Murine CD16/32 (CD16/Fc gamma III and CD32/FcgammaII receptors)
Applications Flow Cytometry