Please login to your online account to display your discounted pricing

Rat anti-CD16/CD32, Clone: 93, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14367201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14367201

  • / Each
 

Description & Specifications

Specifications

Antigen CD16/CD32
Applications Flow Cytometry
Clone 93
Concentration 0.2mg/mL
Conjugate Biotin
Cross Reactivity Mouse
Format Purified
Formulation Phosphate buffer pH 7.2. IgG purified.
Host Species Rat
Immunogen Murine CD16/32 (CD16/Fc gamma III and CD32/FcgammaII receptors)
Isotype IgG2a
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2214
Test Specificity Murine CD16/32 (CD16/Fc gamma III and CD32/FcgammaII receptors)