Please login to your online account to display your discounted pricing

Rat anti-CD117/c-kit, Clone: ACK2, PE, Novus Biologicals™

$197.00 - $418.00

Specifications

Antigen CD117/c-kit
Clone ACK2
Concentration 0.2mg/mL
Immunogen Mouse CD117
Gene ID (Entrez) 3815
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14382101

 
novus biologicals canada
NBP14382101MG
0.1mg Each for $289.00

NBP14382102

 
novus biologicals canada
NBP14382102MG
0.2mg Each for $418.00

NBP14382105

 
novus biologicals canada
NBP143821005MG
0.05mg Each for $197.00
Description & Specifications

Specifications

Antigen CD117/c-kit
Clone ACK2
Concentration 0.2mg/mL
Immunogen Mouse CD117
Gene ID (Entrez) 3815
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2b, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry