Please login to your online account to display your discounted pricing

Rat anti-CD117/c-kit, Clone: ACK2, PE-Cy5, Novus Biologicals™

$197.00 - $461.00

Specifications

Antigen CD117/c-kit
Clone ACK2
Immunogen Mouse CD117
Gene ID (Entrez) 3815
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14307901

 
novus biologicals canada
NBP14307901MG
0.1mg Each for $321.00

NBP14307902

 
novus biologicals canada
NBP14307902MG
0.2mg Each for $461.00

NBP14307905

 
novus biologicals canada
NBP143079005MG
0.05mg Each for $197.00
Description & Specifications

Specifications

Antigen CD117/c-kit
Clone ACK2
Immunogen Mouse CD117
Gene ID (Entrez) 3815
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate PE-Cy5
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry