Please login to your online account to display your discounted pricing

Rat anti-CD117/c-kit, Clone: ACK2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143359005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14335905

  • / Each
 

Description & Specifications

Specifications

Antigen CD117/c-kit
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blot
Clone ACK2
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Gene Accession No. P10721
Host Species Rat
Immunogen Mouse CD117
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3815
Test Specificity The ACK2 monoclonal antibody reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor and stem cell factor receptor.
Target Species Murine