Please login to your online account to display your discounted pricing

Rat anti-CD117/c-kit, Clone: ACK2, APC, Novus Biologicals™

$222.95 - $493.00

Specifications

Antigen CD117/c-kit
Clone ACK2
Concentration 0.2mg/mL
Immunogen Mouse CD117
Gene ID (Entrez) 3815
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14302001

 
novus biologicals canada
NBP14302001MG
0.1mg Each for $321.00

NBP14302002

 
novus biologicals canada
NBP14302002MG
0.2mg Each for $493.00

NBP14302005

 
novus biologicals canada
NBP143020005MG
0.05mg Each for $222.95
Description & Specifications

Specifications

Antigen CD117/c-kit
Clone ACK2
Concentration 0.2mg/mL
Immunogen Mouse CD117
Gene ID (Entrez) 3815
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2b, κ
Conjugate Allophycocyanin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry