Please login to your online account to display your discounted pricing

Rat anti-CD117/c-kit, Clone: 2B8, FITC, Novus Biologicals™

$171.00 - $569.00

Specifications

Antigen CD117/c-kit
Clone 2B8
Concentration 0.5mg/mL
Immunogen Mouse bone marrow mast cells
Gene ID (Entrez) 3815
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14397401

 
novus biologicals canada
NBP14397401MG
0.1mg Each for $246.00

NBP1439745

 
novus biologicals canada
NBP14397405MG
0.5mg Each for $569.00

NBP14397405

 
novus biologicals canada
NBP143974005MG
0.05mg Each for $171.00
Description & Specifications

Specifications

Antigen CD117/c-kit
Clone 2B8
Concentration 0.5mg/mL
Immunogen Mouse bone marrow mast cells
Gene ID (Entrez) 3815
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry