Please login to your online account to display your discounted pricing

Rat anti-CCR7, Clone: 3D12, FITC, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CCR7
Clone 3D12
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CCR7.
Gene Accession No. P32248
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143968

 
novus biologicals canada
NBP143968100TESTS
100 Tests Each for $252.00

NBP14396825

 
novus biologicals canada
NBP14396825TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CCR7
Clone 3D12
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CCR7.
Gene Accession No. P32248
Gene ID (Entrez) 1236
Formulation PBS [pH 7.2] containing BSA and/or gelatin for protein stabilization
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2a, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry