Please login to your online account to display your discounted pricing

Rat anti-CCR7, Clone: 3D12, Biotin, Novus Biologicals™

$131.00 - $296.00

Specifications

Antigen CCR7
Clone 3D12
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CCR7.
Gene Accession No. P32248
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14357001

 
novus biologicals canada
NBP14357001MG
0.1mg Each for $296.00

NBP14357003

 
novus biologicals canada
NBP1435700025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen CCR7
Clone 3D12
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CCR7.
Gene Accession No. P32248
Gene ID (Entrez) 1236
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry