Please login to your online account to display your discounted pricing

Rat anti-CCR7, Clone: 3D12, APC, Novus Biologicals™

$175.00 - $362.00

Specifications

Antigen CCR7
Clone 3D12
Immunogen The immunogen for this antibody was CCR7.
Gene Accession No. P32248
Gene ID (Entrez) 1236
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143011

 
novus biologicals canada
NBP143011100TESTS
100 Tests Each for $362.00

NBP14301125

 
novus biologicals canada
NBP14301125TESTS
25 Tests Each for $175.00
Description & Specifications

Specifications

Antigen CCR7
Clone 3D12
Immunogen The immunogen for this antibody was CCR7.
Gene Accession No. P32248
Gene ID (Entrez) 1236
Formulation PBS [pH 7.2] containing BSA and/or gelatin for protein stabilization
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate APC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry