Please login to your online account to display your discounted pricing

Rat anti-C1q R1/CD93, Clone: AA4.1, Biotin, Novus Biologicals™

$246.00 - $493.00

Specifications

Antigen C1q R1/CD93
Clone AA4.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD93.
Gene ID (Entrez) 22918
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1435255

 
novus biologicals canada
NBP14352505MG
0.5mg Each for $493.00

NBP14352501

 
novus biologicals canada
NBP14352501MG
0.1mg Each for $246.00
Description & Specifications

Specifications

Antigen C1q R1/CD93
Clone AA4.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD93.
Gene ID (Entrez) 22918
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry