Please login to your online account to display your discounted pricing

Rat anti-B7-H3/CD276, Clone: M3.2D7, Biotin, Novus Biologicals™

$171.00 - $386.00

Specifications

Antigen B7-H3/CD276
Clone M3.2D7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Purified CD276.
Gene Accession No. Q5ZPR3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14350801

 
novus biologicals canada
NBP14350801MG
0.1mg Each for $386.00

NBP14350803

 
novus biologicals canada
NBP1435080025MG
0.025mg Each for $171.00
Description & Specifications

Specifications

Antigen B7-H3/CD276
Clone M3.2D7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Purified CD276.
Gene Accession No. Q5ZPR3
Gene ID (Entrez) 80381
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry