Please login to your online account to display your discounted pricing

Rat anti-B7-H2/ICOSLG, Clone: HK5.3, PE, Novus Biologicals™

$354.00 - $493.00

Specifications

Antigen B7-H2/ICOSLG
Clone HK5.3
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was PE B7RP1.
Gene ID (Entrez) 23308
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14373402

 
novus biologicals canada
NBP14373402MG
0.2mg Each for $493.00

NBP14373401

 
novus biologicals canada
NBP14373401MG
0.1mg Each for $354.00
Description & Specifications

Specifications

Antigen B7-H2/ICOSLG
Clone HK5.3
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was PE B7RP1.
Gene ID (Entrez) 23308
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry