missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-B7-H2/ICOSLG, Clone: HK5.3, Biotin, Novus Biologicals™

$280.88 - $280.88

Specifications

Antigen B7-H2/ICOSLG
Clone HK5.3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PE B7RP1.
Gene ID (Entrez) 23308
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14350501 Novus Biologicals Canada
NBP14350501MG
0.1mg Each for $280.88
Specifications Description & Specifications

Specifications

B7-H2/ICOSLG
0.5mg/mL
23308
Monoclonal
Store at 4°C. Do not freeze.
IgG2a, κ
Affinity Purified
Flow Cytometry
HK5.3
The immunogen for this antibody was PE B7RP1.
Phosphate buffer pH 7.2
Primary
Rat
Biotin
Affinity Purified