Please login to your online account to display your discounted pricing

Rat anti-B7-H1/PD-L1/CD274, Clone: 1-111A, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14311701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14311701

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Isotype IgG2a, κ
Applications Flow Cytometry
Conjugate Unlabeled
Quantity 0.1mg
Gene ID (Entrez) 29126
Storage Requirements Store at 4°C. Do not freeze.
Cross Reactivity Mouse
Clone 1-111A
Concentration 0.2mg/mL
Formulation Phosphate buffer pH 7.2
Primary or Secondary Primary
Antigen B7-H1/PD-L1/CD274
Monoclonal or Polyclonal Monoclonal
Host Species Rat
Immunogen The immunogen for this antibody was B7H1.
Purification Method Affinity Purified
Regulatory Status RUO