Please login to your online account to display your discounted pricing

Rat anti-B7-H1/PD-L1/CD274, Clone: 1-111A, Biotin, Novus Biologicals™

$171.00 - $493.00

Specifications

Antigen B7-H1/PD-L1/CD274
Clone 1-111A
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was B7H1.
Gene ID (Entrez) 29126
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14346901

 
novus biologicals canada
NBP14346901MG
0.1mg Each for $246.00

NBP14346905

 
novus biologicals canada
NBP143469005MG
0.05mg Each for $171.00

NBP1434695

 
novus biologicals canada
NBP14346905MG
0.5mg Each for $493.00
Description & Specifications

Specifications

Antigen B7-H1/PD-L1/CD274
Clone 1-111A
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was B7H1.
Gene ID (Entrez) 29126
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry