Please login to your online account to display your discounted pricing

Rat anti-B7-H1/PD-L1/CD274, Clone: 1-111A, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143469005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14346905

  • / Each
 

Description & Specifications

Specifications

Formulation Phosphate buffer pH 7.2
Purification Method Affinity Purified
Quantity 0.05mg
Cross Reactivity Mouse
Antigen B7-H1/PD-L1/CD274
Format Affinity Purified
Host Species Rat
Applications Flow Cytometry
Immunogen The immunogen for this antibody was B7H1.
Isotype IgG2a, κ
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 29126
Clone 1-111A
Concentration 0.5mg/mL
Conjugate Biotin