Please login to your online account to display your discounted pricing

Rat anti-B7-H1/PD-L1/CD274, Clone: 1-111A, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14346901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14346901

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Cross Reactivity Mouse
Quantity 0.1mg
Isotype IgG2a, κ
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Formulation Phosphate buffer pH 7.2
Clone 1-111A
Conjugate Biotin
Regulatory Status RUO
Primary or Secondary Primary
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was B7H1.
Antigen B7-H1/PD-L1/CD274
Host Species Rat
Purification Method Affinity Purified
Gene ID (Entrez) 29126