Please login to your online account to display your discounted pricing

Rat anti-B7-2/CD86, Clone: PO3.1, Novus Biologicals™

$230.00 - $340.00

Specifications

Antigen B7-2/CD86
Clone PO3.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD86.
Gene ID (Entrez) 942
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14338201

 
novus biologicals canada
NBP14338201MG
0.1mg Each for $285.00

NBP14338205

 
novus biologicals canada
NBP143382005MG
0.05mg Each for $230.00

NBP1433825

 
novus biologicals canada
NBP14338205MG
0.5mg Each for $340.00
Description & Specifications

Specifications

Antigen B7-2/CD86
Clone PO3.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD86.
Gene ID (Entrez) 942
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2b, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay