Please login to your online account to display your discounted pricing

Rat anti-B7-2/CD86, Clone: PO3.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14338205MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1433825

  • / Each
 

Description & Specifications

Specifications

Host Species Rat
Cross Reactivity Mouse
Antigen B7-2/CD86
Applications Flow Cytometry
Applications Functional Assay
Isotype IgG2b, κ
Quantity 0.5mg
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 942
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Immunogen The immunogen for this antibody was CD86.
Purification Method Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Clone PO3.1
Concentration 0.5mg/mL
Conjugate Unlabeled