Please login to your online account to display your discounted pricing

Rat anti-B7-2/CD86, Clone: PO3.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14338201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14338201

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Antigen B7-2/CD86
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Applications Flow Cytometry
Applications Functional Assay
Immunogen The immunogen for this antibody was CD86.
Isotype IgG2b, κ
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 942
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Host Species Rat
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Clone PO3.1