Please login to your online account to display your discounted pricing

Rat anti-B7-2/CD86, Clone: PO3.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143382005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14338205

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Cross Reactivity Mouse
Format Affinity Purified
Immunogen The immunogen for this antibody was CD86.
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 942
Antigen B7-2/CD86
Host Species Rat
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 0.05mg
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation Phosphate buffer pH 7.2
Clone PO3.1