Please login to your online account to display your discounted pricing

Rat anti-B7-2/CD86, Clone: GL1, Novus Biologicals™

$219.00 - $340.00

Specifications

Antigen B7-2/CD86
Clone GL1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD86.
Gene ID (Entrez) 942
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1433815

 
novus biologicals canada
NBP14338105MG
0.5mg Each for $340.00

NBP1433815M

 
novus biologicals canada
NBP143381005MG
0.05mg Each for $219.00
Description & Specifications

Specifications

Antigen B7-2/CD86
Clone GL1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD86.
Gene ID (Entrez) 942
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation