Please login to your online account to display your discounted pricing

Rat anti-B7-2/CD86, Clone: GL1, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP143381005MG

 View all options for this product


For Research Use Only

Catalog No. NBP1433815M


Description & Specifications


Antigen B7-2/CD86
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone GL1
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen The immunogen for this antibody was CD86.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 942